1H-Indol-7-ylacetonitrile

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR305244-250MGEA 2580 DKK
APOSOR305244-250MG
1H-Indol-7-ylacetonitrile
1H-Indol-7-ylacetonitrile
Formel: C₁₀H₈N₂
Molekylvægt: 156,184 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09263867
CAS-nummer: 82199-98-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR