1H-Indole-5-carbohydrazide

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR305214-250MGEA 1320 DKK
APOSOR305214-250MG
1H-Indole-5-carbohydrazide
1H-Indole-5-carbohydrazide
Formel: C₉H₉N₃O
Molekylvægt: 175,187 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11977800
CAS-nummer: 406192-82-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR