5-Hydroxy-2-methyl-1-[(4-methylphenyl)sulphonyl]-1H-indole-3-carboxylic acid

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR305195-250MGEA 0 DKK
APOSOR305195-250MG
5-Hydroxy-2-methyl-1-[(4-methylphenyl)sulphonyl]-1H-indole-3-carboxylic acid
5-Hydroxy-2-methyl-1-[(4-methylphenyl)sulphonyl]-1H-indole-3-carboxylic acid
Formel: C₁₇H₁₅NO₅S
Molekylvægt: 345,37 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD23098752
CAS-nummer: 930112-00-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR