(2E)-3-(3H-Imidazo[4,5-b]pyridin-6-yl)prop-2-enoic acid
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR305189-250MGEA
0
DKK
APOSOR305189-250MG
(2E)-3-(3H-Imidazo[4,5-b]pyridin-6-yl)prop-2-enoic acid
(2E)-3-(3H-Imidazo[4,5-b]pyridin-6-yl)prop-2-enoic acid
Formel:
C₉H₇N₃O₂ Molekylvægt: 189,171 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD13177376 CAS-nummer: 335030-65-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...