4-[2-(1H-Imidazol-2-yl)phenyl]pyridine

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR305173-250MGEA 0 DKK
APOSOR305173-250MG
4-[2-(1H-Imidazol-2-yl)phenyl]pyridine
4-[2-(1H-Imidazol-2-yl)phenyl]pyridine
Formel: C₁₄H₁₁N₃
Molekylvægt: 221,257 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13177311
CAS-nummer: 608515-26-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR