N-(Imidazo[1,2-a]pyridin-3-yl)benzamide

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR305154-250MGEA 0 DKK
APOSOR305154-250MG
N-(Imidazo[1,2-a]pyridin-3-yl)benzamide
N-(Imidazo[1,2-a]pyridin-3-yl)benzamide
Formel: C₁₄H₁₁N₃O
Molekylvægt: 237,257 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13177178
CAS-nummer: 860257-98-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR