4-(Piperidin-3-yloxy)-1H-indazole

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR305094-250MGEA 4270 DKK
APOSOR305094-250MG
4-(Piperidin-3-yloxy)-1H-indazole
4-(Piperidin-3-yloxy)-1H-indazole
Formel: C₁₂H₁₅N₃O
Molekylvægt: 217,267 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13183054
CAS-nummer: 478830-50-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR