1,3-Benzothiazole-4-sulphonyl chloride

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR305036-250MGEA 0 DKK
APOSOR305036-250MG
1,3-Benzothiazole-4-sulphonyl chloride
1,3-Benzothiazole-4-sulphonyl chloride
Formel: C₇H₄ClNO₂S₂
Molekylvægt: 233,695 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16251206
CAS-nummer: 149575-65-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR