5-Methyl-1H-benzimidazole-6-boronic acid, pinacol ester, N1-BOC protected

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR305023-250MGEA 3060 DKK
APOSOR305023-250MG
5-Methyl-1H-benzimidazole-6-boronic acid, pinacol ester, N1-BOC protected
5-Methyl-1H-benzimidazole-6-boronic acid, pinacol ester, N1-BOC protected
Formel: C₁₉H₂₇BN₂O₄
Molekylvægt: 358,24 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13182516
CAS-nummer: 631909-46-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR