tert-Butyl 5-methyl-1H-benzimidazole-1-carboxylate

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR305021-250MGEA 0 DKK
APOSOR305021-250MG
tert-Butyl 5-methyl-1H-benzimidazole-1-carboxylate
tert-Butyl 5-methyl-1H-benzimidazole-1-carboxylate
Formel: C₁₃H₁₆N₂O₂
Molekylvægt: 232,278 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13182505
CAS-nummer: 863877-81-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR