N-[3-(Chloromethyl)phenyl]methanesulphonamide

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR305014-1GEA 0 DKK
APOSOR305014-1G
N-[3-(Chloromethyl)phenyl]methanesulphonamide
N-[3-(Chloromethyl)phenyl]methanesulphonamide
Formel: C₈H₁₀ClNO₂S
Molekylvægt: 219,689 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD05663972
CAS-nummer: 362529-31-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR