7-Benzyl-1,7-diazaspiro[3.5]nonane
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR304406-1GEA
11160
DKK
APOSOR304406-1G
7-Benzyl-1,7-diazaspiro[3.5]nonane
7-Benzyl-1,7-diazaspiro[3.5]nonane
Formel:
C₁₄H₂₀N₂ Molekylvægt: 216,322 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD24384637 CAS-nummer: 1420897-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...