3-(Hydroxymethyl)cyclobutan-1-ol

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR304356-1GEA 2240 DKK
APOSOR304356-1G
3-(Hydroxymethyl)cyclobutan-1-ol
3-(Hydroxymethyl)cyclobutan-1-ol
Formel: C₅H₁₀O₂
Molekylvægt: 102,132 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09743755
CAS-nummer: 1245647-03-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR