(2S)-2-(Methoxymethyl)azetidine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR304335-1GEA 6950 DKK
APOSOR304335-1G
(2S)-2-(Methoxymethyl)azetidine
(2S)-2-(Methoxymethyl)azetidine
Formel: C₅H₁₁NO
Molekylvægt: 101,147 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11977228
CAS-nummer: 935668-27-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR