N-[4-(Dimethylsulphamoyl)phenyl]acetamide

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR303917-1GEA 1040 DKK
APOSOR303917-1G APOSOR303917-5G APOSOR303917-500MG
N-[4-(Dimethylsulphamoyl)phenyl]acetamide
N-[4-(Dimethylsulphamoyl)phenyl]acetamide
Formel: C₁₀H₁₄N₂O₃S
Molekylvægt: 242,295 g/mol
Smeltepunkt: 141…143 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01213968
CAS-nummer: 54951-54-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR