2-(Aminomethyl)benzeneboronic acid, pinacol ester hydrochloride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR303550-1GEA 1160 DKK
APOSOR303550-1G APOSOR303550-500MG APOSOR303550-5G
2-(Aminomethyl)benzeneboronic acid, pinacol ester hydrochloride
2-(Aminomethyl)benzeneboronic acid, pinacol ester hydrochloride
Formel: C₁₃H₂₀BNO₂·ClH
Molekylvægt: 269,575 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD24386342
CAS-nummer: 248274-04-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR