[4-(Pyrrolidine-1-carbonyl)phenyl] boronic acid pinacol ester

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR303402-1GEA 0 DKK
APOSOR303402-1G APOSOR303402-5G
[4-(Pyrrolidine-1-carbonyl)phenyl] boronic acid pinacol ester
[4-(Pyrrolidine-1-carbonyl)phenyl] boronic acid pinacol ester
Formel: C₁₇H₂₄BNO₃
Molekylvægt: 301,188 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09027085
CAS-nummer: 1073353-55-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR