3-(1-Benzylpiperidin-4-yl)-3,4-dihydroquinazolin-2-amine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR303335-500MGEA 3390 DKK
APOSOR303335-500MG
3-(1-Benzylpiperidin-4-yl)-3,4-dihydroquinazolin-2-amine
3-(1-Benzylpiperidin-4-yl)-3,4-dihydroquinazolin-2-amine
Formel: C₂₀H₂₄N₄
Molekylvægt: 320,431 g/mol
Smeltepunkt: 102…104 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD25948725
CAS-nummer: 337910-17-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR