N-[(2-Aminophenyl)methyl]-1-benzylpiperidin-4-amine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR303298-500MGEA 1380 DKK
APOSOR303298-500MG
N-[(2-Aminophenyl)methyl]-1-benzylpiperidin-4-amine
N-[(2-Aminophenyl)methyl]-1-benzylpiperidin-4-amine
Formel: C₁₉H₂₅N₃
Molekylvægt: 295,422 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD20257749
CAS-nummer: 79099-03-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR