1-(1-Benzyl-1H-1,2,3-triazol-4-yl)ethan-1-ol

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR303280-500MGEA 985 DKK
APOSOR303280-500MG APOSOR303280-1G
1-(1-Benzyl-1H-1,2,3-triazol-4-yl)ethan-1-ol
1-(1-Benzyl-1H-1,2,3-triazol-4-yl)ethan-1-ol
Formel: C₁₁H₁₃N₃O
Molekylvægt: 203,24 g/mol
Smeltepunkt: 79…81 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD24109296
CAS-nummer: 943432-02-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR