6-Bromo-3-methoxy-2-methylbenzamide

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR303147-500MGEA 0 DKK
APOSOR303147-500MG APOSOR303147-1G
6-Bromo-3-methoxy-2-methylbenzamide
6-Bromo-3-methoxy-2-methylbenzamide
Formel: C₉H₁₀BrNO₂
Molekylvægt: 244,085 g/mol
Smeltepunkt: 108…110 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD22689157
CAS-nummer: 1420537-79-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR