2-(4-(Methylsulphonyl)piperazin-1-yl)thiazol-4-amine hydrochloride

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR302546-1GEA 0 DKK
APOSOR302546-1G
2-(4-(Methylsulphonyl)piperazin-1-yl)thiazol-4-amine hydrochloride
2-(4-(Methylsulphonyl)piperazin-1-yl)thiazol-4-amine hydrochloride
Formel: C₈H₁₄N₄O₂S₂·ClH
Molekylvægt: 298,813 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD26127725
CAS-nummer: 1023815-33-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR