4-Chlorothiazolo[5,4-c]pyridin-2-amine

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR302477-1GEA 0 DKK
APOSOR302477-1G
4-Chlorothiazolo[5,4-c]pyridin-2-amine
4-Chlorothiazolo[5,4-c]pyridin-2-amine
Formel: C₆H₄ClN₃S
Molekylvægt: 185,634 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD22690824
CAS-nummer: 1439824-01-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR