N-(2-Bromopyridin-4-yl)-2,6-dichlorobenzamide

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR302476-1GEA 0 DKK
APOSOR302476-1G
N-(2-Bromopyridin-4-yl)-2,6-dichlorobenzamide
N-(2-Bromopyridin-4-yl)-2,6-dichlorobenzamide
Formel: C₁₂H₇BrCl₂N₂O
Molekylvægt: 346,007 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD22690823
CAS-nummer: 1258298-00-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR