N-(2-Aminopyrimidin-4-yl)acetamide 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR302411-250MGEA 0 DKK
APOSOR302411-250MG
N-(2-Aminopyrimidin-4-yl)acetamide 98%
N-(2-Aminopyrimidin-4-yl)acetamide
Formel: C₆H₈N₄O
Molekylvægt: 152,154 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD22690793
CAS-nummer: 1695410-90-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR