1-Boc-2-(2-Carboxyethyl)piperidine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR302389-1GEA 2730 DKK
APOSOR302389-1G
1-Boc-2-(2-Carboxyethyl)piperidine
1-Boc-2-(2-Carboxyethyl)piperidine
Formel: C₁₃H₂₃NO₄
Molekylvægt: 257,326 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02179009
CAS-nummer: 669713-96-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR