4-Bromo-3-methyl-1-(pyrimidin-2-yl)-1H-pyrazol-5-amine

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR302372-1GEA 0 DKK
APOSOR302372-1G
4-Bromo-3-methyl-1-(pyrimidin-2-yl)-1H-pyrazol-5-amine
4-Bromo-3-methyl-1-(pyrimidin-2-yl)-1H-pyrazol-5-amine
Formel: C₈H₈BrN₅
Molekylvægt: 254,087 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16092775
CAS-nummer: 1250084-93-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR