2,7-Dimethylimidazo[1,2-a]pyridine-3-sulphonyl chloride

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR302246-1GEA 0 DKK
APOSOR302246-1G
2,7-Dimethylimidazo[1,2-a]pyridine-3-sulphonyl chloride
2,7-Dimethylimidazo[1,2-a]pyridine-3-sulphonyl chloride
Formel: C₉H₉ClN₂O₂S
Molekylvægt: 244,698 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD26404012
CAS-nummer: 1335052-39-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR