Tetra-tert-butyl 2,2',2'',2'''-((((2-(tert-butoxy)-2-oxoethyl)azanediyl)bis(ethane-2,1-diyl))bis(azanetriyl))tetraacetate

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR302177-1GEA 0 DKK
APOSOR302177-1G
Tetra-tert-butyl 2,2',2'',2'''-((((2-(tert-butoxy)-2-oxoethyl)azanediyl)bis(ethane-2,1-diyl))bis(azanetriyl))tetraacetate
Tetra-tert-butyl 2,2',2'',2'''-((((2-(tert-butoxy)-2-oxoethyl)azanediyl)bis(ethane-2,1-diyl))bis(azanetriyl))tetraacetate
Formel: C₃₄H₆₃N₃O₁₀
Molekylvægt: 673,878 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD26403989
CAS-nummer: 280563-33-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR