(1-(4-(Dimethylamino)pyridin-2-yl)-1H-pyrazol-3-yl)methanol

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR302138-1GEA 0 DKK
APOSOR302138-1G
(1-(4-(Dimethylamino)pyridin-2-yl)-1H-pyrazol-3-yl)methanol
(1-(4-(Dimethylamino)pyridin-2-yl)-1H-pyrazol-3-yl)methanol
Formel: C₁₁H₁₄N₄O
Molekylvægt: 218,255 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD23704676
CAS-nummer: 1449117-30-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR