4-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR302120-5GEA 0 DKK
APOSOR302120-5G
4-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole
4-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole
Formel: C₈H₁₁IN₂O
Molekylvægt: 278,09 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD19689194
CAS-nummer: 938066-17-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR