4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-acetic acid
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR302119-1GEA
1660
DKK
APOSOR302119-1G
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-acetic acid
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-acetic acid
Formel:
C₁₁H₁₇BN₂O₄ Molekylvægt: 252,075 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD16660234 CAS-nummer: 1083326-41-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...