2-[(Thiophen-2-yl)carbonyl]pyridine

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR302103-1GEA 0 DKK
APOSOR302103-1G
2-[(Thiophen-2-yl)carbonyl]pyridine
2-[(Thiophen-2-yl)carbonyl]pyridine
Formel: C₁₀H₇NOS
Molekylvægt: 189,234 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02260530
CAS-nummer: 6602-63-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR