(1-(2-Morpholinopyridin-4-yl)-1H-pyrazol-4-yl)methanol

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR302069-1GEA 0 DKK
APOSOR302069-1G
(1-(2-Morpholinopyridin-4-yl)-1H-pyrazol-4-yl)methanol
(1-(2-Morpholinopyridin-4-yl)-1H-pyrazol-4-yl)methanol
Formel: C₁₃H₁₆N₄O₂
Molekylvægt: 260,292 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD22690759
CAS-nummer: 1429309-26-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR