1-Allyl-4-nitro-1H-pyrazole

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR302026-1GEA 0 DKK
APOSOR302026-1G
1-Allyl-4-nitro-1H-pyrazole
1-Allyl-4-nitro-1H-pyrazole
Formel: C₆H₇N₃O₂
Molekylvægt: 153,139 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16810706
CAS-nummer: 1240577-26-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR