4-(2-Bromopyridin-4-yl)morpholine

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR302022-1GEA 0 DKK
APOSOR302022-1G
4-(2-Bromopyridin-4-yl)morpholine
4-(2-Bromopyridin-4-yl)morpholine
Formel: C₉H₁₁BrN₂O
Molekylvægt: 243,1 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11869654
CAS-nummer: 1049023-41-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR