5-Chloroimidazo[1,2-a]pyridine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR301061-1GEA 5300 DKK
APOSOR301061-1G
5-Chloroimidazo[1,2-a]pyridine
5-Chloroimidazo[1,2-a]pyridine
Formel: C₇H₅ClN₂
Molekylvægt: 152,581 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD10698083
CAS-nummer: 63111-79-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR