7-Hydroxy-1,2,3,4-tetrahydroisoquinoline

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR301053-1GEA 4540 DKK
APOSOR301053-1G
7-Hydroxy-1,2,3,4-tetrahydroisoquinoline
7-Hydroxy-1,2,3,4-tetrahydroisoquinoline
Formel: C₉H₁₁NO
Molekylvægt: 149,19 g/mol
Smeltepunkt: 179…182 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08234812
CAS-nummer: 30798-64-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR