1-tert-Butylguanidine hydrochloride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR300901-5GEA 0 DKK
APOSOR300901-5G APOSOR300901-1G
1-tert-Butylguanidine hydrochloride
1-tert-Butylguanidine hydrochloride
Formel: C₅H₁₃N₃·ClH
Molekylvægt: 151,638 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD28122847
CAS-nummer: 26536-30-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR