2-Amino-5-bromoquinazolin-4-ol

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR300183-250MGEA 942 DKK
APOSOR300183-250MG
2-Amino-5-bromoquinazolin-4-ol
2-Amino-5-bromoquinazolin-4-ol
Formel: C₈H₆BrN₃O
Molekylvægt: 240,057 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18379108
CAS-nummer: 937668-63-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR