7-Bromo-1,2,3,4-tetrahydro-3-methylisoquinoline

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR300104-500MGEA 0 DKK
APOSOR300104-500MG
7-Bromo-1,2,3,4-tetrahydro-3-methylisoquinoline
7-Bromo-1,2,3,4-tetrahydro-3-methylisoquinoline
Formel: C₁₀H₁₂BrN
Molekylvægt: 226,113 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11559077
CAS-nummer: 848185-12-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR