8-Chloro-3-methyl[1,2,4]triazolo[4,3-a]pyrazine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR300032-1GEA 699 DKK
APOSOR300032-1G
8-Chloro-3-methyl[1,2,4]triazolo[4,3-a]pyrazine
8-Chloro-3-methyl[1,2,4]triazolo[4,3-a]pyrazine
Formel: C₆H₅ClN₄
Molekylvægt: 168,584 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08272087
CAS-nummer: 68774-78-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR