6-Bromo-8-methoxy-[1,2,4]triazolo[4,3-a]pyrazine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR300031-1GEA 8680 DKK
APOSOR300031-1G
6-Bromo-8-methoxy-[1,2,4]triazolo[4,3-a]pyrazine
6-Bromo-8-methoxy-[1,2,4]triazolo[4,3-a]pyrazine
Formel: C₆H₅BrN₄O
Molekylvægt: 229,034 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD20501877
CAS-nummer: 1350885-67-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR