N'-Hydroxy-5-methylisoxazole-3-carboximidamide

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR29712-1GEA 0 DKK
APOSOR29712-1G
N'-Hydroxy-5-methylisoxazole-3-carboximidamide
N'-Hydroxy-5-methylisoxazole-3-carboximidamide
Formel: C₅H₇N₃O₂
Molekylvægt: 141,128 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00210165
CAS-nummer: 90585-88-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR