2-Acetyl-3,5-dimethyl-1H-pyrrole

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR29415-1GEA 0 DKK
APOSOR29415-1G
2-Acetyl-3,5-dimethyl-1H-pyrrole
2-Acetyl-3,5-dimethyl-1H-pyrrole
Formel: C₈H₁₁NO
Molekylvægt: 137,179 g/mol
Smeltepunkt: 115…117 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00982312
CAS-nummer: 1500-93-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR