8-Chlorooct-1-ene 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


APOSOR2941-5GEA 1170 DKK
APOSOR2941-5G
8-Chlorooct-1-ene 97%
8-Chlorooct-1-ene
Formel: C₈H₁₅Cl
Molekylvægt: 146,658 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00671827
CAS-nummer: 871-90-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR