4H,6H-Pyrrolo[1,2-a][4,1]benzoxazepine-4,6-dione

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


Advarsel

APOSOR29335-20MGEA 219 DKK
APOSOR29335-20MG
4H,6H-Pyrrolo[1,2-a][4,1]benzoxazepine-4,6-dione
4H,6H-Pyrrolo[1,2-a][4,1]benzoxazepine-4,6-dione
Formel: C₁₂H₇NO₃
Molekylvægt: 213,189 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00662464
CAS-nummer: 259269-45-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR