2-Methoxychalcone
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSOR27788-1GEA
1460
DKK
APOSOR27788-1G
2-Methoxychalcone
2-Methoxychalcone
Formel:
C₁₆H₁₄O₂ Molekylvægt: 238,281 g/mol Smeltepunkt: 53…57 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00048035 CAS-nummer: 5416-70-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...