trans-3-Ethoxycinnamic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR27554-100MGEA 0 DKK
APOSOR27554-100MG APOSOR27554-1G
trans-3-Ethoxycinnamic acid
trans-3-Ethoxycinnamic acid
Formel: C₁₁H₁₂O₃
Molekylvægt: 192,211 g/mol
Smeltepunkt: 131…134 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00016843
CAS-nummer: 188545-72-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR