3-Hydroxy-7-methyl-1,2-benzisoxazole 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR27361-1GEA 0 DKK
APOSOR27361-1G APOSOR27361-50MG
3-Hydroxy-7-methyl-1,2-benzisoxazole 97%
3-Hydroxy-7-methyl-1,2-benzisoxazole
Formel: C₈H₇NO₂
Molekylvægt: 149,147 g/mol
Smeltepunkt: 155…157 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00104669
CAS-nummer: 36238-83-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR